Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen symbol:CACNG2
- Clonality:Monoclonal
- Clone:S245-36
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:203 to 323
- Storage buffer:PBS pH 7,4, 50% glycerol
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Fusion protein amino acids 203-323 (Cytoplasmic Cterminus) of rat TARPGamma2,
- Purification:purified by Protein G chromatography
- Size:100µg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1?g/ml
Type: Primary
Antigen: Cacng2
Clonality: Monoclonal
Clone: S245-36
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2A
Reactivity: Mouse, Rat