Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen symbol:MAP2K3
- Clonality:Monoclonal
- Clone:6B12
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Monkey,Human,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:39.1 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinan protein of human MAP2K3(NP_659731) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: MAP2K3
Clonality: Monoclonal
Clone: 6B12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat