Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-ASCC1 Mouse Monoclonal Antibody [clone: 2D7]
Anti-ASCC1 Mouse Monoclonal Antibody [clone: 2D7]
Catalog # ORIGCF503668
Supplier:  OriGene
Anti-ASCC1 Mouse Monoclonal Antibody [clone: 2D7]
Catalog # ORIGCF503668
Supplier:  OriGene

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    activating signal cointegrator 1 complex subunit 1
  • Antigen symbol:
    ASCC1
  • Clonality:
    Monoclonal
  • Clone:
    2D7
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    Full length
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    41 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Full length human recombinant protein of human ASCC1(NP_057031) produced in HEK293T cell.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: ASCC1
Clonality: Monoclonal
Clone: 2D7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human