Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen symbol:Xcl1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:1 to 92
- Storage buffer:100 µg (0,5 mg/ml) Protein A purified rabbit anti-mouse Lymphotactin polyclonal antibody in phosphate buffered saline (PBS), pH 7,2, containing 50% glycerol, 1% BSA, 0,02% thimerosal.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:E. coli-expressed mouse lymphotactin (a.a. 1-92)
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Western Blot: 0.5-4 ?g/ml; Immunoprecipitation: 4-8?g/ml
Type: Primary
Antigen: Xcl1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse