Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-Fbxo32 Rabbit Polyclonal Antibody
  ORIGTA318919
 :  OriGene
Anti-Fbxo32 Rabbit Polyclonal Antibody
  ORIGTA318919
 :  OriGene

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen symbol:
    Fbxo32
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    100 µg (0,5 mg/ml) affinity purified rabbit anti-Fbx32 polyclonal antibody in phosphate buffered saline (PBS), pH 7,2, containing 30% glycerol, 0,5% BSA, 5 mM EDTA and 0,01% thimerosal.
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Synthetic peptide surrounding amino acid 56 of rat Fbx32
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

 

 

Recommended Dilutions: Western Blot: 1-4 ?g/ml

Type: Primary
Antigen: Fbxo32
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat