Anti-Protein A Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BSBTBA1120-1
Supplier: Boster Bio
Specifications
- Antibody type:Tag
- Antigen name:Protein A
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Cross adsorption:No
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Tag
Antigen: Protein A
Clonality:
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host:
Isotype:
Reactivity: