Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:Succinate dehydrogenase complex, subunit A, flavoprotein
- Antigen symbol:SDHA
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Gene ID:6389
- Antigen synonyms:PGL5|CMD1GG|FP|SDHF|SDH1|SDH2
- UniProtKB:P31040
- Storage temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Immunogen:E.coli-derived human SDHA recombinant protein (Position: S44-L380). Human SDHA shares 98.2% and 97.6% amino acid (aa) sequence identity with mouse and rat SDHA, respectively.
- Purification:Immunogen affinity purified.
- Size:100 µg/vial
- Pk:100 µG
Rabbit IgG polyclonal antibody for Succinate dehydrogenase [ubiquinone] flavoprotein subunit, mitochondrial(SDHA) detection. Tested with WB in Human;Mouse;Rat.
Complex II of the mitochondrial respiratory chain, also known as succinate dehydrogenase or succinate:ubiquinone oxidoreductase, consists of 4 nuclear-encoded polypeptides, these are the flavoprotein subunit (SDHA), the iron sulfur protein subunit (SDHB), and the integral membrane protein subunits SDHC and SDHD. SDHA is an acronym for succinate dehydrogenase complex subunit A. The succinate dehydrogenase (SDH) protein complex catalyzes the oxidation of succinate (succinate + ubiquinone => fumarate + ubiquinol). The SDHA subunit is connected to the SDHB subunit on the hydrophilic, catalytic end of the complex, and weighs 72.7 kDA. Mutations in the SDHA subunit have a distinct pathology from mutations in the SDHB/SDHC/SDHD subunits; it is the only subunit to never have shown tumor suppressor behaviour. Heterozygous carriers of an SDHA mutation do not develop paragangliomas as has been seen for mutations in the other subunits. This appears to be due to the expression of two similar SDHA genes (Types I and II) in the paraganglia system.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: SGK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human