Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-RETN Rabbit Polyclonal Antibody
Anti-RETN Rabbit Polyclonal Antibody
Catalog # BSBTPB9412
Supplier:  Boster Bio
undefined
Anti-RETN Rabbit Polyclonal Antibody
Catalog # BSBTPB9412
Supplier:  Boster Bio

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Resistin
  • Antigen symbol:
    RETN
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Gene ID:
    56729
  • Antigen synonyms:
    FIZZ3|RETN1|RSTN|XCP1|ADSF
  • UniProtKB:
    Q9HD89
  • Storage temperature:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Immunogen:
    E. coli-derived human Resistin recombinant protein (Position: K19-P108). Human Resistin shares 58.6% amino acid (aa) sequence identity with mouse Resistin.
  • Purification:
    Immunogen affinity purified.
  • Size:
    100 µg/vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Resistin(RETN) detection. Tested with WB, ELISA in Human.

Resistin, also known as adipose tissue-specific secretory factor (ADSF) or C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein (XCP1) is a cysteine-rich protein that in humans is encoded by the RETN gene. The resistin gene comprises 4 exons, the first of which is untranslated, and spans approximately 1,750 bp. The human resistin gene is localized to a cloned fragment of human chromosome 19. In primates, pigs and dogs, resistin is secreted by immune and epithelial cells while in rodents, it is secreted by adipose tissue. Resistin is a cytokine whose physiologic role has been the subject of much controversy regarding its involvement with obesity and type II diabetes mellitus.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

WB: The detection limit for Resistin is approximately 0.25ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: TNFRSF11A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: Yes
Host: Rabbit
Isotype: IgG
Reactivity: Mouse