Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:adrenoceptor beta 2, surface
- Antigen symbol:ADRB2
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Lyophilized
- Gene ID:154
- Antigen synonyms:BETA2AR|ADRBR|BAR|ADRB2R|B2AR
- UniProtKB:P07550
- Storage temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human ADRB2(221-256aa RVFQEAKRQLQKIDKSEGRFHVQNLSQVEQDGRTGH), different from the related mouse and rat sequences by three amino acids.
- Purification:Immunogen affinity purified.
- Size:100 µg/vial
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Beta-2 adrenergic receptor(ADRB2) detection. Tested with WB in Human;Rat.
ADRB2, also known as beta-2 adrenergic receptor, is a beta-adrenergic receptor within a cell membrane which reacts with adrenaline (epinephrine) as a hormone or neurotransmitter affecting muscles or organs. It is mapped to 5q32. This receptor is directly associated with one of its ultimate effectors, the class C L-type calcium channel Ca(V)1.2. The genetic variation in the ADRB2 gene may be of major importance for obesity, energy expenditure, and lipolytic ADRB2 function in adipose tissue, at least in women. What’s more, it has been found that activation of ADRB2 receptors can stimulate gamma-secretase activity and beta-amyloid production, and the ADRB2 receptors activator may contribute to beta-amyloid accumulation in AD.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
WB: The detection limit for ADRB2 is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: AIFM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat