Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-HSD17B10 Rabbit Polyclonal Antibody
Anti-HSD17B10 Rabbit Polyclonal Antibody
Catalog # BSBTPB9336
Supplier:  Boster Bio
undefined
Anti-HSD17B10 Rabbit Polyclonal Antibody
Catalog # BSBTPB9336
Supplier:  Boster Bio

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    hydroxysteroid (17-beta) dehydrogenase 10
  • Antigen symbol:
    HSD17B10
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Gene ID:
    3028
  • Antigen synonyms:
    MRX31|ERAB|17b-HSD10|MRX17|HADH2|MRPP2|HCD2|MRXS10|MHBD|SCHAD|CAMR|ABAD|SDR5C1|DUPXp11.22
  • UniProtKB:
    Q99714
  • Storage temperature:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Immunogen:
    E.coli-derived human ERAB recombinant protein (Position: E48-P261). Human ERAB shares 87% and 88% amino acid (aa) sequence identity with mouse and rat ERAB, respectively.
  • Purification:
    Immunogen affinity purified.
  • Size:
    100 µg/vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for 3-hydroxyacyl-CoA dehydrogenase type-2(HSD17B10) detection. Tested with WB, IHC-P in Human;Mouse.

ERAB is also known as HSD17B10 or HADH2. This gene encodes 3-hydroxyacyl-CoA dehydrogenase type II, a member of the short-chain dehydrogenase/reductase superfamily. The gene product is a mitochondrial protein that catalyzes the oxidation of a wide variety of fatty acids and steroids, and is a subunit of mitochondrial ribonuclease P, which is involved in tRNA maturation. The protein has been implicated in the development of Alzheimer disease, and mutations in the gene are the cause of 17beta-hydroxysteroid dehydrogenase type 10 (HSD10) deficiency. Several alternatively spliced transcript variants have been identified, but the full-length nature of only two transcript variants has been determined.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

WB: The detection limit for ERAB is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: HSPD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat