Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:caspase 8
- Antigen symbol:CASP8
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Form:Lyophilized
- Gene ID:12370
- Antigen synonyms:FLICE|Mch5|MACH|CASP-8
- UniProtKB:O89110
- Storage temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Immunogen:E.coli-derived mouse Caspase-8 recombinant protein (Position: S388-P480). Mouse Caspase-8 shares 83% amino acid (aa) sequence identity with human Caspase-8.
- Purification:Immunogen affinity purified.
- Size:100 µg/vial
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Caspase-8(CASP8) detection. Tested with WB in Mouse.
CASP8 is also known as CAP4, MACH or MCH5. This gene encodes a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes composed of a prodomain, a large protease subunit, and a small protease subunit. Activation of caspases requires proteolytic processing at conserved internal aspartic residues to generate a heterodimeric enzyme consisting of the large and small subunits. This protein is involved in the programmed cell death induced by Fas and various apoptotic stimuli. The N-terminal FADD-like death effector domain of this protein suggests that it may interact with Fas-interacting protein FADD. This protein was detected in the insoluble fraction of the affected brain region from Huntington disease patients but not in those from normal controls, which implicated the role in neurodegenerative diseases. Many alternatively spliced transcript variants encoding different isoforms have been described, although not all variants have had their full-length sequences determined.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
WB: The detection limit for Caspase-8 is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: CASP9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human