Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CD47 Rabbit Antibody
Anti-CD47 Rabbit Antibody
Catalog # BSBTPA2223
Supplier:  Boster Bio
undefined
Anti-CD47 Rabbit Antibody
Catalog # BSBTPA2223
Supplier:  Boster Bio

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD47 molecule
  • Antigen symbol:
    CD47
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    IAP|MER6|OA3
  • Amino acid number:
    308 to 323
  • Sequence:
    EPLNAFKESKGMMNDE
  • Storage temperature:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human CD47 (308-323aa EPLNAFKESKGMMNDE), identical to the related rat sequence.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: CD47
Clonality:
Clone:
Conjugation:
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat