Scintillation vials, without caps, KIMBLE®
Catalog # KIML74506-20
Supplier: KIMBLEKONTES
Specifications
- Cap included:No
- Capacity:22,00 ml
- Pk:500
Specifications
About this item
These vials made from KG-33 borosilicate glass are packaged in cellular trays which have alphanumeric coordinates for fast, easy identification.
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Packaging: Each tray contains 100 vials. Five trays per case, shrink-wrapped to preserve cleanliness.