Order Entry
Northern Ireland
ContactUsLinkComponent
Hexadecyl palmitate, EMPROVE® ESSENTIAL Ph. Eur., NF, SAFC®
Hexadecyl palmitate, EMPROVE® ESSENTIAL Ph. Eur., NF, SAFC®
Catalog # 1.15419.9024
CAS Number:  
Hexadecyl palmitate, EMPROVE® ESSENTIAL Ph. Eur., NF, SAFC®
Catalog # 1.15419.9024
CAS Number:  

Specifications

  • Pk:
    20 kg
  • Seq:
    4241

Specifications

About this item

 :
 :480.86 g/mol
Storage Temperature:Ambient
EINECS:208-736-6
CAS Number:540-10-3
Formula:C₃₂H₆₄O₂
MDL Number:MFCD00053739