Order Entry
Northern Ireland
ContactUsLinkComponent
Drying cabinets, insulated, E3
Drying cabinets, insulated, E3
Supplier:  GENLAB
Certificates
undefined
Drying cabinets, insulated, E3
Supplier:  GENLAB

Specifications

  • Catalog No:
  • 466-0464
  • 466-0462
  • 466-0467
  • 466-0461
  • Model:
  • E3DWC425/N
  • E3DWC425/F
  • E3DWC885/F
  • E3DWC200/N
  • Capacity:
  • 425 L
  • 425 L
  • 885 L
  • 200 L
  • Convection type:
  • Natural convection
  • Forced convection
  • Forced convection
  • Natural convection
  • Shelves (shelf positions):
  • 3 (12)
  • 3 (12)
  • 3 (12)
  • 3 (4)
  • Int. W×D×H:
  • 530×590×1350 mm
  • 530×590×1350 mm
  • 1110×590×1350 mm
  • 930×450×490 mm
  • Ext. W×D×H:
  • 600×650×1755 mm
  • 600×650×1755 mm
  • 1180×650×1755 mm
  • 1000×500×660 mm
  • Energy consumption:
  • 13.27* kWh/day
  • 18.32* kWh/day
  • 27.18* kWh/day
  • 8.65* kWh/day
  • Power consumption:
  • 1750 W
  • 1750 W
  • 2500 W
  • 750 W

Specifications

Frequently Bought Together