Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Furniture
Storage Cabinets
Accessories for storage cabinets for solvents
Accessories for storage cabinets for solvents
Accessories for storage cabinets for solvents
Catalog # LBCP9910000
Supplier:
LABCONCO
Expand image
Accessories for storage cabinets for solvents
Catalog # LBCP9910000
Supplier:
LABCONCO
Specifications
Description:
Solvent vent kit consisting metal flexible hose Ø 38 mm, two hose clamps and slip fit outlet adapter
Pk:
1
Specifications
About this item
Product Details
Product Details