Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Hamster
- Western blot:Yes
- Cross adsorption:No
- Format:Purified IgG
- Form:Clear, fluorescent yellow liquid
- Storage buffer:10 mM Sodium phosphate, 0,15 M Sodium chloride, pH 7,2, 1 % (w/v) BSA, Protease/IgG free
- Storage temperature:2 - 8 °C
- Concentration:1,50 mg/ml (E 1% at 280 nm = 13,0)
- Shipping temperature:2 - 8 °C
- Immunogen:Hamster IgG
- Tested applications:Fluoresence ELISA, Fluorescence-Activated Cell Sorting
- Purification:Affinity purified using solid phaseHamster IgG
- Size:2,0 mg
- Pk:2 mg
ImmunoReagents’ FITC Conjugates are used in immunofluorescence microscopy, flow cytometry, FLISA (fluorescent ELISA) and high throughput screening assays. Fluorescein is typically excited by the 488 nm line of an argon laser, and emission is collected at 518 nm. FITC antibody conjugates are yellow-orange in color but emit a yellow-green color upon Em 518 nm. FITC antibody conjugates are sensitive to light. The typical FITC to antibody ratio (F/P) is >4.0 Excitation/Emission = 494nm / 518nm Emission Color = Green (Similar Dyes: Alexa Fluor 488, Cy2, DyLight ® 488)
- Origin: US
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hamster