Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CKMM from skeletal muscle Goat Polyclonal Antibody
Anti-CKMM from skeletal muscle Goat Polyclonal Antibody
Catalog # IMMRIR1416
Supplier:  ImmunoReagents
Anti-CKMM from skeletal muscle Goat Polyclonal Antibody
Catalog # IMMRIR1416
Supplier:  ImmunoReagents

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Creatine Kinase MM
  • Antigen symbol:
    CKMM
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Liquid, 0,2 µm filtered
  • Antigen synonyms:
    Purified Human CKMM from skeletal muscle
  • Storage buffer:
    10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2
  • Storage temperature:
    2...8 °C
  • Concentration:
    >10 mg/ml (E 1% at 280 nm = 1,3)
  • Shipping temperature:
    2...8 °C
  • Immunogen:
    Human CKMM from skeletal muscle
  • Tested applications:
    Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling
  • Purification:
    Proprietary 2-step fractionation procedure
  • Size:
    1,0 ml
  • Pk:
    1 ml

Specifications

About this item

ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.  

  • Origin: US

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002.
Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Primary
Antigen: CKMM from skeletal muscle
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human