Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Alpha-1-Antichymotrypsin Goat Polyclonal Antibody
Anti-Alpha-1-Antichymotrypsin Goat Polyclonal Antibody
Catalog # IMMRIR1368
Supplier:  ImmunoReagents
Anti-Alpha-1-Antichymotrypsin Goat Polyclonal Antibody
Catalog # IMMRIR1368
Supplier:  ImmunoReagents

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Alpha-1-Antichymotrypsin
  • Antigen symbol:
    ACT
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Clear, colorless liquid, 0,2 µm filtered
  • Antigen synonyms:
    MGC88254|GIG24|GIG25
  • Storage buffer:
    10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2
  • Storage temperature:
    2...8 °C
  • Concentration:
    >4,5 mg/ml (E 1% at 280 nm = 13,0)
  • Shipping temperature:
    2...8 °C
  • Immunogen:
    Human Alpha-1-Antichymotrypsin
  • Tested applications:
    Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling
  • Purification:
    Affinity purified using solid phase Human Alpha-1-Antichymotrypsin
  • Size:
    1,0 mg
  • Pk:
    1 mg

Specifications

About this item

ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.  

  • Origin: US

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Type: Primary
Antigen: Alpha-1-Antichymotrypsin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human