Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Tissue Plasminogen Activator Mouse Monoclonal Antibody
Anti-Tissue Plasminogen Activator Mouse Monoclonal Antibody
Catalog # USBIT5600-11B
Supplier:  US Biological
Anti-Tissue Plasminogen Activator Mouse Monoclonal Antibody
Catalog # USBIT5600-11B
Supplier:  US Biological

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tissue Plasminogen Activator
  • Antigen symbol:
    TPA
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    0,05M sodium phosphate, 0,15M sodium chloride, pH 7,4.
  • Storage temperature:
    Cool
  • Concentration:
    ~2 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Type: Primary
Antigen: Tissue Plasminogen Activator
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse