Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Tissue Plasminogen Activator Rabbit Polyclonal Antibody
Anti-Tissue Plasminogen Activator Rabbit Polyclonal Antibody
Catalog # USBIT5600-05C
Supplier:  US Biological
Anti-Tissue Plasminogen Activator Rabbit Polyclonal Antibody
Catalog # USBIT5600-05C
Supplier:  US Biological

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tissue Plasminogen Activator
  • Antigen symbol:
    TPA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    0,05M sodium phosphate, 0,1M sodium chloride, 1mM EDTA, pH 6.6.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Tissue Plasminogen Activator
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human