Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Small Cell Lung Carcinoma, Neural and Endocrine Antigen Mouse Monoclonal Antibody [clone: MOC-21]
Anti-Small Cell Lung Carcinoma, Neural and Endocrine Antigen Mouse Monoclonal Antibody [clone: MOC-21]
Catalog # USBIS1014-57N
Supplier:  US Biological
Anti-Small Cell Lung Carcinoma, Neural and Endocrine Antigen Mouse Monoclonal Antibody [clone: MOC-21]
Catalog # USBIS1014-57N
Supplier:  US Biological

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Small Cell Lung Carcinoma, Neural and Endocrine Antigen
  • Clonality:
    Monoclonal
  • Clone:
    MOC-21
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Western blot:
    Yes
  • Format:
    Supernatant
  • Form:
    liquid
  • Storage buffer:
    0,1% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: Small Cell Lung Carcinoma, Neural and Endocrine Antigen
Clonality: Monoclonal
Clone: MOC-21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: