Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:Protein Kinase C Iota
- Antigen symbol:PRKCI
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Modification:phospho-Thr555, phospho-Thr563
- Storage buffer:PBS(without Mg2+ and Ca2+), pH 7,3, 1mg/mlBSA (IgG, protease free), 0,05% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:10 Pieces
- Pk:10
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: Protein Kinase C, iota, lambda
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human