Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-Protein Kinase C, lambda Rabbit Polyclonal Antibody
  USBIP9103-22L
Anti-Protein Kinase C, lambda Rabbit Polyclonal Antibody
  USBIP9103-22L

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase C Iota
  • Antigen symbol:
    PRKCI
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,25 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Western Blot: 1-3?g/ml

Type: Primary
Antigen: Protein Kinase C, lambda
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat