Anti-Platelet Basic Protein Rabbit Polyclonal Antibody
Product Details & Documents
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100
Type: Primary
Antigen: Platelet Basic Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Documents
Specifications
- Size:200 µl
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by ammonium sulfate precipitation
- Antibody type:Primary
- Epitope:N-terminal
- Form:liquid
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Shipping temperature:Ice
- Antigen name:Platelet Basic Protein
- Cat. No.:USBIP0023-01F
- Reactivity:Human
- Conjugation:Unconjugated
- Storage buffer:PBS, 0,09% sodium azide.
- ELISA:Yes
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



