Anti-19-Nortestosterone Sheep Polyclonal Antibody
Catalog # USBIN4004
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:19-Nortestosterone
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- Form:liquid
- Storage buffer:20mM PBS, 0,09% sodium azide, pH 7,2.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: ELISA:10 ?g/ml
Type: Primary
Antigen: 19-Nortestosterone
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: