Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:Myosin Light Chain 2
- Antigen symbol:MYL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS(without Mg2+ and Ca2+), pH 7,4, 150mMsodium chloride, 0,02% sodium azide and 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 1:500-1:1000
Type: Primary
Antigen: Myosin Light Chain 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse