Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-lambda light chain Goat Polyclonal Antibody
Catalog # USBIL1185-11L
Supplier:  US Biological
Anti-lambda light chain Goat Polyclonal Antibody
Catalog # USBIL1185-11L
Supplier:  US Biological

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    lambda light chain
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Format:
    Serum
  • Form:
    liquid
  • Storage buffer:
    TBS, pH 7,4, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Turbidimetry
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Type: Secondary
Antigen: LAMBDA LIGHT CHAIN
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human