Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Junctional Adhesion Molecule 1 Goat Polyclonal Antibody
Anti-Junctional Adhesion Molecule 1 Goat Polyclonal Antibody
Catalog # USBIJ8009-05C
Supplier:  US Biological
Anti-Junctional Adhesion Molecule 1 Goat Polyclonal Antibody
Catalog # USBIJ8009-05C
Supplier:  US Biological

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Junctional Adhesion Molecule 1
  • Antigen symbol:
    JAM1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Form:
    liquid
  • Storage buffer:
    Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: ELISA: 1:2000

Type: Primary
Antigen: Junctional Adhesion Molecule 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Human