Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Junctional Adhesion Molecule 1
- Antigen symbol:JAM1
- Clonality:Monoclonal
- Clone:10B1403
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:250 µg
- Pk:250 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:100-1:200;
Type: Primary
Antigen: Junctional Adhesion Molecule 1
Clonality: monoclonal
Clone: 10B1403
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse