Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-Glucose Regulated Protein 78 Rabbit Polyclonal Antibody
  USBIG3057-20A
Anti-Glucose Regulated Protein 78 Rabbit Polyclonal Antibody
  USBIG3057-20A

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen name:
    Glucose Regulated Protein 78
  • Antigen symbol:
    GRP78
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Hamster,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    643 to 654
  • Storage buffer:
    0,05% sodium azide, before the addition ofglycerol to 40 .
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Western Blot: 2?g/ml

Type: Primary
Antigen: Glucose Regulated Protein 78
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Mouse, Rat