Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Endoplasmic Reticulum Protein 72 Rabbit polyclonal antibody
Anti-Endoplasmic Reticulum Protein 72 Rabbit polyclonal antibody
Catalog # USBIE2291-77C
Supplier:  US Biological
Anti-Endoplasmic Reticulum Protein 72 Rabbit polyclonal antibody
Catalog # USBIE2291-77C
Supplier:  US Biological

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Endoplasmic Reticulum Protein 72
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Monkey,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    10 mM sodium HEPES (pH 7,5), 150 mM NaCl, 100ug/ml BSA and 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: Endoplasmic Reticulum Protein 72
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat