Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CRISP3 Mouse monoclonal antibody [clone: 9E88]
Anti-CRISP3 Mouse monoclonal antibody [clone: 9E88]
Catalog # USBIC9018-58B
Supplier:  US Biological
Anti-CRISP3 Mouse monoclonal antibody [clone: 9E88]
Catalog # USBIC9018-58B
Supplier:  US Biological

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cysteine-rich Secretory Protein 3
  • Antigen symbol:
    CRISP3
  • Clonality:
    Monoclonal
  • Clone:
    9E88
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    Cool
  • Concentration:
    ~2 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: ELISA: 1:200-1:400

Type: Primary
Antigen: CRISP3 (cysteine-rich secretory protein 3)
Clonality: Monoclonal
Clone: 9E+88
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human