Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:Chlamydia trachomatis, psittaci
- Clonality:Monoclonal
- Clone:11C207
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG3
- Form:liquid
- Storage buffer:10mMsodium phosphate,150mM sodium chloride, pH 7,4, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: Chlamydia trachomatis, psittaci
Clonality: Monoclonal
Clone: 11C207
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: