Anti-EPCAM Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: 158206]
USBI145455
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:Epithelial cell adhesion molecule
- Antigen symbol:EPCAM
- Clonality:Monoclonal
- Clone:158206
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, 0,5% BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 Tests
- Pk:100 Tests
Type: Primary
Antigen: EPCAM (epithelial cell adhesion molecule)
Clonality: monoclonal
Clone: 158206
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human