Anti-POLE2 Mouse Monoclonal Antibody [clone: 1A3]
Catalog # USBI131567
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Polymerase (DNA directed), epsilon 2, accessory subunit
- Antigen symbol:POLE2
- Clonality:Monoclonal
- Clone:1A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 101
- Storage buffer:PBS, pH 7,2.
- Sequence:MAPERLRSRALSAFKLRGLLLRGEAIKYLTEALQSISELELEDKLEKIINAVEKQPLSSNMIERSVVEAAVQECSQSVDETIEHVFNIIGAFDIPRFVYN
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: POLE2 (polymerase (DNA directed), epsilon 2, accessory subunit)
Clonality: monoclonal
Clone: 1A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat