Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-ISL1 Mouse Monoclonal Antibody [clone: 1A3]
Anti-ISL1 Mouse Monoclonal Antibody [clone: 1A3]
Catalog # USBI128608
Supplier:  US Biological
Anti-ISL1 Mouse Monoclonal Antibody [clone: 1A3]
Catalog # USBI128608
Supplier:  US Biological

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ISL LIM homeobox 1
  • Antigen symbol:
    ISL1
  • Clonality:
    Monoclonal
  • Clone:
    1A3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    63 to 159
  • Storage buffer:
    PBS, pH 7,2.
  • Sequence:
    YCKRDYIRLYGIKCAKCSIGFSKNDFVMRARSKVYHIECFRCVACSRQLIPGDEFALREDGLFCRADHDVVERASLGAGDPLSPLHPARPLQMAAEP
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: ISL1 (ISL LIM homeobox 1)
Clonality: monoclonal
Clone: 1A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human