Anti-PTPRC Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 30-F11]
Catalog # ROCK200-526-N83
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase receptor type C
- Antigen symbol:PTPRC
- Clonality:Monoclonal
- Clone:30-F11
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Storage buffer:0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 0,1% Gelatin
- Storage temperature:Cool
- Concentration:0,2 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Anti-CD45 Antibody (Monoclonal) was produced by repeated immunizations with CD45 antigen.
- Tested applications:Fluoresence ELISA
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-CD45 is useful for Flow Cytometry, Immunoprecipitation, immunochemistry, Immunofluorescence, and Western Blot using mouse spleen cells, or an appropriate cell type (where indicated).
Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze!
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 30-F11
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse