Anti-ITGAM Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: M1/70]
Catalog # ROCK200-508-N79
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:integrin, Alpha M (complement component 3 receptor 3 subunit)
- Antigen symbol:ITGAM
- Clonality:Monoclonal
- Clone:M1/70
- Conjugation:RPE (R-Phycoerythrin)
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Storage buffer:0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:Sufficient to run approximately 100 tests
- Shipping temperature:Ice
- Immunogen:Anti-CD11b Antibody (Monoclonal) was produced by repeated immunizations with CD11b antigen.
- Tested applications:Fluoresence ELISA
- Purification:affinity-purified
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Anti-CD11b is useful for Immunoprecipitation, immunochemistry, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.
Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze! Light sensitive!
Type: Primary
Antigen: ITGAM
Clonality: Monoclonal
Clone: M1/70
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse