Anti-CD8A Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 53-6.7]
Catalog # ROCK200-508-N77
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:CD8 antigen, alpha chain
- Antigen symbol:CD8A
- Clonality:Monoclonal
- Clone:53-6.7
- Conjugation:RPE (R-Phycoerythrin)
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:12525
- Antigen synonyms:BB154331|Ly-2|Lyt-2|Ly-35|Ly-B
- Storage buffer:0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:Sufficient to run approximately 100 tests
- Shipping temperature:Ice
- Immunogen:Anti-CD8a Antibody (Monoclonal) was produced by repeated immunizations with Mouse thymus or spleen .
- Tested applications:Fluoresence ELISA
- Purification:affinity-purified
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Anti-CD8a is useful for immunochemistry, Immunoprecipitation, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.
Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze! Light sensitive!
Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse