Anti-Kappa Light Chainkappa light chain Rabbit Polyclonal Antibody (Biotin)
ROCK610-4610
- Antibody type:Secondary
- Antigen name:Kappa Light Chain
- Antigen symbol:IGKC
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:none
- ChIP:Yes
- Neutralisation:Yes
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Mouse kappa light chain
- Purification:purified by immunoaffinity chromatography
- Size:1 mg
- Pk:1 mg
Secondary Rabbit Anti-κ (kappa chain) Reacts with Mouse
Recommended Dilutions: ELISA: 1:600,000; Western Blot: 1:10000 - 1:50000; Immunochemistry: 1:2000 - 1:10000;
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: Igκ
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Light Chain
Host: Rabbit
Isotype: IgG
Reactivity: Mouse