Anti-GFP Mouse Polyclonal Antibody (Unconjugated) [clone: 9F9.F9]
Catalog # ROCK600-301-215
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Tag
- Antigen name:Green Fluorescence Protein
- Antigen symbol:GFP
- Clonality:Monoclonal
- Clone:9F9.F9
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Multiple
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- ChIP:Yes
- Neutralisation:Yes
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Recombinant Green Fluorescent Protein (GFP) fusion Protein corresponding to the full length amino acid sequence (246 aa) derived from the jellyfish Aequorea victoria.
- Purification:affinity-purified
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Monoclonal anti-GFP is designed to detect enhanced GFP and GFP containing recombinant Proteins. This antibody can be used to detect GFP by ELISA (sandwich or capture) for the direct binding of antigen.
Recommended Dilutions: ELISA: 1:80,000 - 1:500,000; Western Blot: 1:3000 - 1:30,000; Immunochemistry: 1:1000 - 1:5000;
Type: Tag
Antigen: GFP
Clonality: Polyclonal
Clone: 9F9.F9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1 kappa
Reactivity: pan