Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Avidin Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Anti-Avidin Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Catalog # ROCK200-4297
Anti-Avidin Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Catalog # ROCK200-4297

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Avidin
  • Antigen symbol:
    AVD
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Multiple
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    20 mg
  • Pk:
    20 mg

Specifications

About this item

Type: Primary
Antigen: Avidin
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: pan