Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-MPO Rabbit Polyclonal Antibody
Anti-MPO Rabbit Polyclonal Antibody
Catalog # ROCK200-4181
undefined
Anti-MPO Rabbit Polyclonal Antibody
Catalog # ROCK200-4181

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Myeloperoxidase
  • Antigen symbol:
    MPO
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Gene ID:
    4353
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Gentamicin Sulfate.
  • Storage temperature:
    Cool
  • Concentration:
    5,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Myeloperoxidase (Human Leukocytes)
  • Purification:
    multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above
  • Size:
    500 µg
  • Pk:
    500 µG

Specifications

About this item

This antibody is suitable for western blotting, ELISA and immunoprecipitation.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 100 µl
Do not add Sodium Azide!

Type: Primary
Antigen: MPO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human