Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-EpCAM-1 (Ab-1) (VU-1D9) Mouse Monoclonal Antibody [clone: VU-1D9]
Anti-EpCAM-1 (Ab-1) (VU-1D9) Mouse Monoclonal Antibody [clone: VU-1D9]
Catalog # OP187-100
Anti-EpCAM-1 (Ab-1) (VU-1D9) Mouse Monoclonal Antibody [clone: VU-1D9]
Catalog # OP187-100

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    EpCAM-1
  • Clonality:
    Monoclonal
  • Clone:
    VU-1D9
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: EpCAM-1 (Ab-1) (VU-1D9)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human