Anti-c-ErbB2/c-Neu (Ab-4) (7.16.4) Mouse Monoclonal Antibody [clone: 7.16.4]
OP16-100
- Antibody type:Primary
- Antigen name:V-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2
- Antigen symbol:C-ERBB2
- Clonality:Monoclonal
- Clone:7.16.4
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: c-ErbB2/c-Neu (Ab-4) (7.16.4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: