Anti-MSH2 (Ab-2) (FE11) Mouse Monoclonal Antibody [clone: FE11]
Catalog # NA27-100
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen symbol:MSH2
- Clonality:Monoclonal
- Clone:FE11
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: MSH2 (Ab-2) (FE11)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: