Anti-MMP-2 (Ab-4) (75-7F7) Mouse Monoclonal Antibody [clone: 75-7F7]
IM51-100
- Antibody type:Primary
- Antigen symbol:MMP-2
- Clonality:Monoclonal
- Clone:75-7F7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: MMP-2 (Ab-4) (75-7F7)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: