Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-Phosphotyrosine (PY20) Mouse Monoclonal Antibody [clone: PY20]
  525295-100
Anti-Phosphotyrosine (PY20) Mouse Monoclonal Antibody [clone: PY20]
  525295-100

 

  • Antibody type:
    Primary
  • Antigen name:
    Phosphotyrosine
  • Antigen symbol:
    PY
  • Clonality:
    Monoclonal
  • Clone:
    PY20
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
    Multiple
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: Phosphotyrosine (PY20)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Pan