Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-ACTB Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
  GENSA00730-200
 :  Genscript
undefined
Anti-ACTB Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
  GENSA00730-200
 :  Genscript

 

  • Antibody type:
    Tag
  • Antigen name:
    Actin beta
  • Antigen symbol:
    ACTB
  • Clonality:
    Monoclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Western blot:
    Yes
  • Antigen synonyms:
    PS1TP5-binding protein 1|cytoplasmic 1|PS1TP5BP1|BRWS1|beta cytoskeletal actin|actin
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    5x40 µg
  • Pk:
    200 µG

 

 

Type: Tag
Antigen: ACTB (actin, beta)
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: